missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Histone Deacetylase Polyclonal Antibody
GREENER_CHOICE

Product Code. 17781214
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17781214 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17781214 Supplier Invitrogen™ Supplier No. PA5119816

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive Control: Rice tissue lysates.

Histone deacetylases (HDACs) are enzymes that remove acetyl groups from lysine residues in the NH2 terminal tails of core histones, resulting in a more closed chromatin structure and repression of gene expression.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone Deacetylase
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Host Species Rabbit
Immunogen Recombinant protein within rice Histone deacetylase aa 150-352.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.