Learn More
Novus Biologicals™ Histone H1 Antibody (derlotuximab) - Chimeric, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27977-50ug
 View more versions of this product
Product Code. 30045798
  • € 213.00 / 50µg
Description
Histone H1 Monoclonal antibody specifically detects Histone H1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H1
Monoclonal
Unconjugated
Q92522
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
derlotuximab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
H1 histone family, member 1, H1.1, H1A, H1F1HIST1, histone 1, H1a, histone cluster 1, H1a, histone H1.1, MGC126642, MGC138345
Histone H1
50 μg
Cellular Markers, Chromatin Research, Epigenetics, Nuclear Envelope Markers
3024