Learn More
Novus Biologicals™ Histone H1 Antibody (derlotuximab) - Chimeric, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-27977-1mg
 View more versions of this product
Product Code. 30048638
  • € 920.00 / 1mg
Description
Histone H1 Monoclonal antibody specifically detects Histone H1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H1
Monoclonal
Unconjugated
Q92522
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
derlotuximab
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
H1 histone family, member 1, H1.1, H1A, H1F1HIST1, histone 1, H1a, histone cluster 1, H1a, histone H1.1, MGC126642, MGC138345
Histone H1
1 mg
Cellular Markers, Chromatin Research, Epigenetics, Nuclear Envelope Markers
3024