missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1 Mouse, Clone: AE-4, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 3005-MSM1-P1
This item is not returnable.
View return policy
Specifications
| Histone H1 | |
| Nuclear | |
| AE-4 | |
| Unconjugated | |
| NP_005309 | |
| Mouse | |
| ∽30kDa | |
| Protein A/G Purified | |
| RUO | |
| 3005 | |
| IgG2a κ |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 1mM PBS with 0.05% BSA and 0.05% azide. | |
| H1 | |
| Nuclei of human leukemia biopsy cells | |
| HeLa, A-431, LNCap or Jurkat cells. Breast carcinoma. | |
| 100 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction