missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18201-1-AP-150UL
This item is not returnable.
View return policy
Description
This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.Specifications
| Histone H1 | |
| Polyclonal | |
| Unconjugated | |
| HIST1H1A | |
| H1.1, H1F1, HIST1, HIST1H1A, histone cluster 1, H1a, Histone H1.1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291145, 3024, 80838 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| D4A3K5, P43275, Q02539 | |
| HIST1H1A | |
| Histone H1 Fusion Protein Ag12704 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction