Learn More
Histone H1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18201-1-AP-20UL
 View more versions of this product
Product Code. 16898254
  • € 145.00 / 20µL
Description
This locus is the ortholog of a mouse protein-coding gene that displays characteristics of a linker histone and is expressed in nuclei of late maturing spermatids. The human locus is expressed; however, the open reading frame has been disrupted by a frameshift so it is unlikely to encode a functional protein.
Specifications
Specifications
Histone H1
Polyclonal
Unconjugated
HIST1H1A
H1.1, H1F1, HIST1, HIST1H1A, histone cluster 1, H1a, Histone H1.1
Rabbit
Antigen Affinity Chromatography
RUO
291145, 3024, 80838
-20°C
Liquid
Immunoprecipitation, Western Blot
0.29 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
D4A3K5, P43275, Q02539
HIST1H1A
Histone H1 Fusion Protein Ag12704
20 μL
Primary
Mouse, Human, Rat