Learn More
Histone H1 Rabbit, Clone: HH1/1784R, enQuireBio™
Rabbit Monoclonal Antibody
Brand: EnquireBio 3005-RBM4-P0
This item is not returnable.
View return policy
Specifications
| Histone H1 | |
| Nuclear | |
| HH1/1784R | |
| Unconjugated | |
| NP_005309 | |
| Rabbit | |
| ∽30kDa | |
| Protein A/G Purified | |
| RUO | |
| 3005 | |
| IgG κ |
| Immunohistochemistry | |
| Monoclonal | |
| 200μg/mL | |
| 10mM PBS with or without 0.05% BSA and 0.05% azide. | |
| H1 | |
| Recombinant full-length human Histone H1 protein | |
| HeLa, A-431, LNCap or Jurkat cells. Breast carcinoma. | |
| 20 μg | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |