missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2a, p Ser129 Antibody (SR1183), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21526-100ul
This item is not returnable.
View return policy
Description
Histone H2a Monoclonal antibody specifically detects Histone H2a in Yeast samples. It is validated for Western Blot
Specifications
| Histone H2a | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Yeast | |
| Purified |
| Western Blot | |
| SR1183 | |
| Western Blot 1:1000-1:2000 | |
| H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460 | |
| A synthesized peptide derived from yeast phospho-Histone H2a [Ser129] (Uniprot #: P04912) | |
| 100 μg | |
| Chromatin Research, Epigenetics | |
| 8338 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction