Learn More
Histone H2A.Z [ac Lys4] Antibody (RM221) [Janelia Fluor™ 669], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals (Bio-Techne) NBP3-26025JF669
Product Code. 30178292
  • € 790.00 / 0.10mL
Description
Histone H2A.Z Monoclonal antibody specifically detects Histone H2A.Z in Vertebrate samples. It is validated for ELISA,Western Blot,Multiplex Immunoassay,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H2A.Z
Monoclonal
Janelia Fluor 669
H2A histone family, member Z, H2A.z, H2A/z, H2AZ histone, H2AZMGC117173, histone H2A.Z
An acetyl-peptide corresponding to Histone H2A.Z [ac Lys4]
0.1 mL
Epigenetics
3015
Store at 4°C in the dark.
IgG
ELISA, Western Blot, Multiplex, Immunocytochemistry/Immunofluorescence
RM221
50mM Sodium Borate
Rabbit
Protein A purified
RUO
Primary
Vertebrates