missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2A.Z, ac Lys7 Antibody (RM222) - Azide and BSA Free, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26026
This item is not returnable.
View return policy
Description
Histone H2A.Z Monoclonal antibody specifically detects Histone H2A.Z in Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
Specifications
| Histone H2A.Z | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Vertebrates | |
| Purified |
| Western Blot, ELISA, Immunocytochemistry, Multiplex | |
| RM222 | |
| Western Blot : 0.5 ug/ml - 2 ug/ml, ELISA : 0.2 ug/ml - 1 ug/ml, Immunocytochemistry/ Immunofluorescence : 1 ug/ml - 2 ug/ml, Multiplex Immunoassay : 0.05 ug/ml - 0.5 ug/ml | |
| H2A histone family, member Z, H2A.z, H2A/z, H2AZ histone, H2AZMGC117173, histone H2A.Z | |
| An acetyl-peptide corresponding to Histone H2A.Z [ac Lys7] | |
| 100 μg | |
| Epigenetics | |
| 3015 | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction