missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AE [p Ser1] Antibody (RM216) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26017JF635
This item is not returnable.
View return policy
Description
Histone H2AE Monoclonal antibody specifically detects Histone H2AE in Human,Vertebrate samples. It is validated for ELISA,Western Blot,Multiplex Immunoassay,Immunocytochemistry/ Immunofluorescence
Specifications
| Histone H2AE | |
| Monoclonal | |
| Janelia Fluor 635 | |
| H2A histone family, member M, H2A/m, H2AFA, H2AFMHistone H2A/m, HIST1H2AE, histone 1, H2ab, histone cluster 1, H2ab, histone H2A type 1-B/E, Histone H2A.2, Histone H2A/a | |
| An unmodified peptide corresponding to the N-terminus of Histone H2AE | |
| 0.1 mL | |
| Epigenetics | |
| 3012 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot, Multiplex, Immunocytochemistry/Immunofluorescence | |
| RM216 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction