missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2AX, p Ser139 Antibody (2207D), Alexa Fluor™ 700, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB2288N
This item is not returnable.
View return policy
Description
Histone H2AX Monoclonal antibody specifically detects Histone H2AX in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| Histone H2AX | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 2207D | |
| Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| H2A.X, H2A/X, H2AFX | |
| Phospho-peptide containing human Histone H2AX S139 site, Accession # P16104 | |
| 0.1 mL | |
| Checkpoint signaling, DNA Double Strand Break Repair, DNA Repair, Epigenetics, Mitotic Regulators, Phospho Specific | |
| 3014 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction