missing translation for 'onlineSavingsMsg'
Learn More

Histone H2AY/macroH2A.1 Antibody (CL5256) [mFluor Violet 500 SE], Novus Biologicals™

Product Code. 30524580 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30524580 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30524580 Supplier Novus Biologicals Supplier No. NBP343837MFV500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Histone H2AY/macroH2A.1 Monoclonal antibody specifically detects Histone H2AY/macroH2A.1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2AY/macroH2A.1
Applications Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CL5256
Conjugate mFluor Violet 500 SE
Formulation 50mM Sodium Borate
Gene Alias core histone macro-H2A.1, H2A histone family, member Y, H2A.y, H2A/y, H2AF12M, H2AFJ, Histone H2A.y, Histone macroH2A1, histone macroH2A1.1, histone macroH2A1.2, MACROH2A1, MACROH2A1.1, macroH2A1.2, Medulloblastoma antigen MU-MB-50.205, mH2A1
Host Species Mouse
Immunogen This antibody was generated using a synthetic peptide of O75367, with the exact immunogen sequence remaining proprietary.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9555
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.