missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B, ac Lys11 Antibody (RM456), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26035H
This item is not returnable.
View return policy
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Multiplex Immunoassay
Specifications
| Histone H2B | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
| Western Blot, ELISA, Multiplex | |
| RM456 | |
| Western Blot, ELISA, Multiplex Immunoassay | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| An acetyl-peptide corresponding to Histone H2B [Lys11] (Uniprot # P62807) | |
| 0.1 mL | |
| Chromatin Research, Epigenetics | |
| 440689 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction