missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Antibody (RM230) [Janelia Fluor™ 525], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26043JF525
This item is not returnable.
View return policy
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Vertebrate samples. It is validated for ELISA,Western Blot,Multiplex Immunoassay,Immunocytochemistry/ Immunofluorescence
Specifications
| Histone H2B | |
| Monoclonal | |
| Janelia Fluor 525 | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| A peptide corresponding to the C-terminus of Human Histone H2B | |
| 0.1 mL | |
| Chromatin Research, Epigenetics | |
| 440689 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, Western Blot, Multiplex, Immunocytochemistry/Immunofluorescence | |
| RM230 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Vertebrates | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction