missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B Antibody (SR1182), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | Histone H2B |
|---|---|
| Clone | SR1182 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681014
|
Novus Biologicals
NBP3-21669-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18648854
|
Novus Biologicals
NBP3-21669-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Histone H2B Monoclonal antibody specifically detects Histone H2B in Yeast samples. It is validated for Western BlotSpecifications
| Histone H2B | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Yeast | |
| FLJ35099, FLJ56780, FLJ56787, histone 2, H2bf, histone cluster 2, H2bf, histone H2B type 2-F, MGC131639 | |
| A synthesized peptide derived from yeast Histone H2B (Uniprot #: P02293) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1182 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Chromatin Research, Epigenetics | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 440689 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title