missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H2B (K123ub) monoclonal antibody, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15846.50ug
This item is not returnable.
View return policy
Specifications
| Histone H2B (K123ub) | |
| Monoclonal | |
| Western Blot (1:500) ChIP (2 ug/CHIP) The optimal working dilution should be determined by the end user. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot | |
| Unconjugated | |
| In PBS (0.02% sodium azide). | |
| A synthetic peptide (conjugated with KLH) corresponding to yeast histone H2B, ubiquitinylated at lysine 123. | |
| RUO | |
| Human, Yeast | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction