missing translation for 'onlineSavingsMsg'
Learn More

Histone H2B, member S Antibody, Novus Biologicals™

Product Code. 18781694 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
Unit Size:
0.1mg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18781694 0.1 mg 0.1mg
18064684 0.025 mg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18781694 Supplier Novus Biologicals Supplier No. NB10056347

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 13 publications

Histone H2B, member S Polyclonal specifically detects Histone H2B, member S in Human, Mouse, Rat, Canine, Chicken, Drosophila, Mammal, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunoblotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H2B, member S
Applications Western Blot, Immunohistochemistry (Paraffin), Immunoblot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 2 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 10 ug/ml, Immunoblotting reported in scientific literature (PMID 28235947)
Formulation PBS with 0.05% Sodium Azide
Gene Alias H2B histone family S member10H2B/s, H2B histone family, member S, Histone H2B.s
Gene Symbols H2BFS
Host Species Rabbit
Immunogen This antibody was generated by immunizing rabbits with a synthetic peptide corresponding to amino acid residues 111-125 AVSEGTKAVTKYTSS of human Histone H2B; GenBank no. CAI19747.1.
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Chromatin Research
Primary or Secondary Primary
Gene ID (Entrez) 54145
Test Specificity A 15 kDa band is observed. This antibody is routinely quality controlled in human lysates.
Target Species Human, Mouse, Rat, Canine, Chicken, Drosophila, Mammalia, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.