Learn More
Histone H2B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27740-1-AP-20UL
 View more versions of this product
Product Code. 18026405
  • € 145.00 / 20µL
Description
The Histone H2B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Histone H2B. This antibody recognizes human antigen. The Histone H2B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
Histone H2B
350 μg/mL
Rabbit
Primary
Human
150 kDa
IgG
Western Blot, ELISA
H2B clustered histone 26, H2B type 12, H2B3B, H2BC26, HIST3H2BB, histone cluster 3, H2bb, Histone H2B type 3 B
20 μL
128312
Store at -20°C. Stable for one year after shipment.