Learn More
Histone H2B Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27740-1-AP-20UL
Description
The Histone H2B antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Histone H2B. This antibody recognizes human antigen. The Histone H2B antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| Histone H2B | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| H2B clustered histone 26, H2B type 12, H2B3B, H2BC26, HIST3H2BB, histone cluster 3, H2bb, Histone H2B type 3 B | |
| 20 μL | |
| 128312 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |