missing translation for 'onlineSavingsMsg'
Learn More

Histone H3.1, Monomethyl Arg17 Antibody (3E10), Novus Biologicals™

Product Code. 30034040 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μL
50 μL
Unit Size:
100µL
50µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30034040 100 μL 100µL
30028840 50 μL 50µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30034040 Supplier Novus Biologicals Supplier No. NBP326131100ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Histone H3.1 Monoclonal antibody specifically detects Histone H3.1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen Histone H3.1
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 3E10
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA, Immunocytochemistry/Immunofluorescence 1:50-1:500
Formulation PBS, pH 7.4, 150mM NaCl and 50% glycerol
Gene Alias H3 histone family, member D, H3.1HIST1H3D, H3/d, H3FA, H3FB, H3FC, H3FDHIST1H3F, H3FF, H3FH, H3FI, H3FJ, H3FK, H3FL, HIST1H3A, HIST1H3B, HIST1H3C, HIST1H3G, HIST1H3H, HIST1H3I, HIST1H3J, histone 1, H3e, histone cluster 1, H3e, histone H3.1, Histone H3/a, Histone H3/b, Histone H3/c, Histone H3/d, Histone H3/f, Histone H3/h, Histone H3/i, Histone H3/j, Histone H3/k, Histone H3/l
Host Species Rabbit
Immunogen Synthesized peptide [UniProt P68431]
Purification Method Affinity purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Epigenetics, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 8353
Target Species Human
Content And Storage Store at -20 to -70C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.