Learn More
Histone H3.3A Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98768MFV500
Product Code. 18596013
  • € 428.00 / 0.10mL
Description
Histone H3.3A Polyclonal antibody specifically detects Histone H3.3A in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Histone H3.3A.
0.1 mL
Chromatin Research, Epigenetics, Phospho Specific
Store at 4C in the dark.