Learn More
Histone H3 Antibody (1B1-B2), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-36468F
Product Code. 18629545
  • ONLINE EXCLUSIVE PRICE
    € 702.00
    € 663.60 / 0.10mL
    Save € 38.40
    5% Off
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), ChIP assay (ChIP), Single Cell Western
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H3
Monoclonal
FITC
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
This Histone H3 antibody (1B1-B2) was raised against synthetic peptide corresponding to the C-terminus of human Histone H3.
0.1 mL
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
126961
Store at 4°C in the dark.
IgG3 κ
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), ChIP Assay
1B1-B2
PBS
Mouse
Protein G purified
RUO
Primary
Human, Mouse, Rat