Learn More
Histone H3(dimethyl K18) Rabbit, Clone: RM168, Abnova™
Rabbit monoclonal antibody raised against synthetic dimethyl peptide of human Histone H3 (dimethyl K18).
Brand: Abnova MAB12778.100ug
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6p22-p21.3. [provided by RefSeq]
Specifications
| Histone H3(dimethyl K18) | |
| Monoclonal | |
| Unconjugated | |
| Dot Blot ELISA (0.1 ug/mL-0.5 ug/mL) Western Blot (0.2 ug/mL-1 ug/mL) The optimal working dilution should be determined by the end user. | |
| H3/A/H3FA | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 8350 | |
| Human | |
| Liquid |
| Dot Blot, ELISA, Western Blot | |
| RM168 | |
| Rabbit monoclonal antibody raised against synthetic dimethyl peptide of human Histone H3 (dimethyl K18). | |
| In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
| HIST1H3A | |
| A synthetic dimethyl peptide corresponding to residues surrounding K18 of Histone H3. | |
| 100 μg | |
| Primary | |
| This antibody reacts to Histone H3 dimethylated at Lysine 18. No cross reactivity with monomethylated Lysine 18 or trimethylated Lysine 18, or other methylation in histone H3. | |
| Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.