Learn More
Histone H3, p Ser10 Antibody (RM163), FITC, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-26018F
Product Code. 30027981
  • € 872.00 / 0.10mL
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H3
Monoclonal
FITC
PBS
Rabbit
Protein A purified
RUO
Primary
Human, Vertebrates
Purified
Western Blot, ELISA, Immunocytochemistry, Multiplex
RM163
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783
This Histone H3 [p Ser10] antibody (RM163) was raised against a phospho-peptide corresponding to Histone H3 [p Ser10]
0.1 mL
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
126961
Store at 4C in the dark.