missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, p Ser28 Antibody (117C826), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-56595MFV610
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Fish samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Histone H3 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Fish | |
| Purified |
| Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 117C826 | |
| Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| This Histone H3 [p Ser28] antibody (117C826) was raised against a synthetic peptide containing phospho serine at position 28 CARKS*APATGGVKK of human Histone H3. The immunogen sequence is conserved across mammalian and invertebrates, as well as plants. | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction