missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3, p Thr6 Antibody (RM160), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26020AF532
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Multiplex Immunoassay
Specifications
| Histone H3 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
| Western Blot, ELISA, Multiplex | |
| RM160 | |
| Western Blot, ELISA, Multiplex Immunoassay | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| This Histone H3 [p Thr6] antibody (RM160) was raised against a phospho-peptide corresponding to Histone H3 [p Thr6] | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction