missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3 pan monoclonal antibody, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB15826.50ug
This item is not returnable.
View return policy
Specifications
| HIST1H3A | |
| Monoclonal | |
| Western Blot (1:1000) ChIP (1-2 ug/CHIP) Immunofluorescence (1:500) The optimal working dilution should be determined by the end user. | |
| histone cluster 1, H3a | |
| HIST1H3A | |
| A synthetic peptide (conjugated with KLH) corresponding to unmodified sequence at C-terminus of Histone H3 pan. | |
| RUO | |
| 8350 | |
| Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
| ChIP Assay, Immunofluorescence, Western Blot | |
| Unconjugated | |
| In PBS (0.05% sodium azide). | |
| H3/A/H3FA | |
| Mouse | |
| 50 μg | |
| Primary | |
| Human, Mouse, Plant | |
| Antibody | |
| IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction