missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H3 [Trimethyl Lys36] Antibody (RM491) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-26038JF635
This item is not returnable.
View return policy
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human,Vertebrate samples. It is validated for ELISA,ChIP assay (ChIP),Multiplex Immunoassay
Specifications
| Histone H3 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
| A trimethyl-peptide corresponding to Histone H3 [Lys36]. (Uniprot # P84243) | |
| 0.1 mL | |
| Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
| 126961 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA, ChIP Assay, Multiplex | |
| RM491 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction