missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4, ac Lys5 Antibody, Alexa Fluor™ 405, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB21-2024AF405
This item is not returnable.
View return policy
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse, Rat, Porcine, Mammal samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| Histone H4 (ac Lys5) | |
| Polyclonal | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| P62805 | |
| HIST4H4 | |
| Synthetic aceylated peptide surrounding Lysine 5 of human Histone H4 [Swiss Prot P62805]. | |
| 0.05 mL | |
| Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones | |
| 121504 | |
| Human, Mouse, Rat, Pig, Mammalia | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, mouse and rat. Predicted to react with most mammal species. | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction