Learn More
Histone H4 Antibody (Immunomedics patent anti-Histone H4), HRP, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals NBP3-28031H
Product Code. 30115820
  • € 431.00 / 0.10mL
Description
Histone H4 Monoclonal antibody specifically detects Histone H4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H4
Monoclonal
HRP
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Histone H4
0.1 mL
Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific
121504
Store at 4°C in the dark.
IgG1
Flow Cytometry, ELISA, Functional Assay
Immunomedics patent anti-Histone H4
PBS
Human
Protein A purified