missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4, Monomethyl Arg3 Antibody (RM195), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-61556
This item is not returnable.
View return policy
Description
Histone H4 Monoclonal antibody specifically detects Histone H4 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
Specifications
| Histone H4 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 0.5 μg/mL-2 μg/mL, ELISA 0.2 μg/mL - 1 μg/mL, Immunocytochemistry/ Immunofluorescence 1 μg/mL - 2 μg/mL, Multiplex Immunoassay 0.2 μg/mL - 1 μg/mL | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| A monomethyl -peptide corresponding to the Histone H4 [Monomethyl Arg3](RM195) | |
| 100 μg | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific | |
| 121504 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunofluorescence, Multiplex | |
| RM195 | |
| Unconjugated | |
| 50% Glycerol/PBS, 1% BSA | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction