missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Histone H4, Monomethyl Lys5 Antibody (RM457), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26036AF350
This item is not returnable.
View return policy
Description
Histone H4 Monoclonal antibody specifically detects Histone H4 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Multiplex Immunoassay
Specifications
| Histone H4 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Vertebrates | |
| Purified |
| Western Blot, ELISA, Multiplex | |
| RM457 | |
| Western Blot, ELISA, Multiplex Immunoassay | |
| H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
| A monomethyl-peptide corresponding to Histone H4 [Lys5] (Uniprot # P62805) | |
| 0.1 mL | |
| Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific | |
| 121504 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction