Learn More
Histone H4, p Ser1 Antibody (RM194), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-61553
Product Code. 18660487
  • € 519.00 / 100µg
Description
Histone H4 Monoclonal antibody specifically detects Histone H4 in Human, Vertebrate samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Multiplex Immunoassay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone H4
Monoclonal
1 mg/mL
Western Blot 0.5 μg/mL-2 μg/mL, ELISA 0.2 μg/mL - 1 μg/mL, Immunocytochemistry/ Immunofluorescence 1 μg/mL - 2 μg/mL, Multiplex Immunoassay 0.1 μg/mL - 1 μg/mL
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
A phospho-peptide corresponding to Histone H4 [p Ser1]
100 μg
Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Phospho Specific
121504
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA, Immunofluorescence, Multiplex
RM194
Unconjugated
50% Glycerol/PBS, 1% BSA
Rabbit
Protein A purified
RUO
Primary
Human, Vertebrates