Learn More
Invitrogen™ Histone Macro-H2A.1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5115532
Product Code. 17248313
  • € 497.00 / 100µL
Description
Antibody detects endogenous levels of total macroH2A.1.
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Histone Macro-H2A.1
Polyclonal
Unconjugated
MACROH2A1
Core histone macro-H2A.1; H2A histone family member Y; H2A histone family, member Y; H2A.y; H2A/y; H2AF12M; H2AFY; Histone H2A.y; histone macroH2A1; histone macroH2A1.1; histone macroH2A1.2; macroH2A1; MACROH2A1.1; macroH2A1.2; medulloblastoma antigen MU-MB-50.205; mH2A1
Rabbit
Affinity chromatography
RUO
26914, 29384, 9555
-20°C
Liquid
Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide
O75367, Q02874, Q9QZQ8
MACROH2A1
A synthesized peptide derived from human H2AFY(Accession O75367), corresponding to amino acid residues K292-M342.
100 μL
Primary
Human, Mouse, Rat