missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 Gag p24 Antibody (9B6C2B7), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-06452CL1
This item is not returnable.
View return policy
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for ELISA,Western Blot
Specifications
| HIV-1 Gag p24 | |
| Monoclonal | |
| CoraFluor 1 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| ELISA, Western Blot | |
| 9B6C2B7 | |
| PBS | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HIV-1 Gag p24 (Accession#: ACI05538.1; Pro 133-Leu 363). | |
| 0.1 mL | |
| Infections (Virus Bacteria and Parasites) | |
| 155030 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction