missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals HIV-1 Gag p24 Antibody (HIV1-24/661) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-34636-0.1mg
This item is not returnable.
View return policy
Description
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| HIV-1 Gag p24 | |
| Monoclonal | |
| 1.0 mg/mL | |
| ELISA 1-5μg/mL for coating, Immunocytochemistry/ Immunofluorescence 0.5-1 μg/mL | |
| P04585, P04585 | |
| Recombinant HIV-1 Gag p24 protein (Uniprot: P04585) | |
| 0.1 mg | |
| Infections (Virus Bacteria and Parasites) | |
| 155030 | |
| Store at -20°C to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| ELISA, Immunocytochemistry | |
| HIV1-24/661 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction