missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 gp120 Antibody (4F4E4), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06629AF700
This item is not returnable.
View return policy
Description
HIV-1 gp120 Monoclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| HIV-1 gp120 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 4F4E4 | |
| Western Blot, ELISA | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant HIV-1 gp120 (Ala 29-Glu 503; 97CN001 strain). | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| 155971 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction