missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 gp120 Antibody, Biotin, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NB100-65513
This item is not returnable.
View return policy
Description
HIV-1 gp120 Polyclonal specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA.
Specifications
| HIV-1 gp120 | |
| Polyclonal | |
| Biotin | |
| PBS (pH 7.2) with 0.05% Sodium Azide | |
| env | |
| Purified gp120 from strain IIIB | |
| 0.5 mL | |
| Virology Bacteria and Parasites | |
| 155971 | |
| Virus | |
| Purified |
| Western Blot, ELISA | |
| 4.0mg/mL | |
| Western Blot 1:100-1:2000, ELISA 1:100-1:2000 | |
| Envelope surface glycoprotein gp120, gp120, gp120 glycoprotein, Human immunodeficiency virus 1, SU, Surface protein | |
| Goat | |
| IgG purified | |
| RUO | |
| Primary | |
| NB100-66437 is specific for gp120, an exterior envelope glycoprotein on the surface of the virus particle that acts as a receptor for CD4 and chemokine receptors on macrophages and T-lymphocytes. It does not react with human T cells, B cells, normal bovine serum or normal human serum. In western blotting a predominant gp120 band is observed. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction