Learn More
HIV-1 gp120 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81822MFV500
This item is not returnable.
View return policy
Description
HIV-1 gp120 Polyclonal antibody specifically detects HIV-1 gp120 in Virus samples. It is validated for Western Blot, ELISA
Specifications
| HIV-1 gp120 | |
| Polyclonal | |
| HIV-1 gp120 antibody was raised against a peptide corresponding to 15 amino acids near the center of HIV-1 gp120. Amino Acid Squence: VAPTKAKRRVVQREKR | |
| 0.1 mL |
| Western Blot | |
| Rabbit | |
| Peptide affinity purified | |
| RUO |