missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HIV-1 Tat specific factor 1 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-18912DL594
This item is not returnable.
View return policy
Description
HIV-1 Tat specific factor 1 Polyclonal antibody specifically detects HIV-1 Tat specific factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| HIV-1 Tat specific factor 1 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| cofactor required for Tat activation of HIV-1 transcription, dJ196E23.2, HIV TAT specific factor 1, HIV Tat-specific factor 1, HIV-1 Tat specific factor 1, TAT-SF1 | |
| The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human HIV-1 Tat specific factor 1 using the numbering given in entry NP_055315.2 (GeneID 27336). | |
| 0.1 mL | |
| Immunology | |
| 27336 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction