missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIV1 nef Monoclonal Antibody (N5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA171507
This item is not returnable.
View return policy
Description
This antibody recognizes HIV-1 subtypes A, B and C, and has been mapped to amino acids 62-68 (EEEVGFP - B subtype consensus). This clone may also be referred to as N5 (01-011).
HIV-1 Nef is one of the accessory proteins synthesized in the early stage of AIDS virus reproduction and is abundantly found in infected cells. The name derives from its negative factor thought at the beginning but presently it is remarked as the protein which bears a most distinctive biological characteristic of AIDS virus. The protein interacts directly with the signal transduction protein of the host T cell and works effectively on AIDS infection or on long term survival of the infected cells or induces apoptosis of non-infected cells. It is also involved in the endocytosis and degradation of receptor protein of the cell surface such as CD 4 and MH4, important for AIDS virus infection.
Specifications
| HIV1 nef | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Virus | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot | |
| N5 | |
| Unconjugated | |
| HIV-1 Nef | |
| Recombinant Nef protein of HIV-1 B-subtype consensus purified from E. coli. | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction