missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HIV1 p24 Monoclonal Antibody (12G4)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA544992

Product Code. 17858632

  • € 494.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Application notes: Sandwich ELISA.

HIV-1 Gag p24 is a capsid protein that constitutes the core of the AIDS virus HIV-1 and is produced by the digestion of its precursor Gag p55 by HIV-1 protease. The HIV-1 Gag p24 protein is indispensable to the reproduction of AIDS virus, and constitutes an essential element for the AIDS virus particle construction. HIV-1 Gag p24 is detectable from the early stage of AIDS virus infection, and is used as a marker for observation of the development in the patient's condition after treatment, as it indicated the amount of virus in the blood.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HIV1 p24
Monoclonal
2 mg/mL
PBS with no preservative; pH 7.4
Mouse
Protein G
RUO
Virus
Antibody
IgG1
ELISA, Western Blot
12G4
Unconjugated
HIV p24; HIV1gp1
Recombinant protein within HIV1 p24 protein aa 133-363 (Catalog#HA210278).
100 μL
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.