missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ HIVEP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567799
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
HIVEP2 is a member of a family of closely related, large, zinc finger-containing transcription factors. It regulates transcription by binding to regulatory regions of various cellular and viral genes that maybe involved in growth, development and metastasis.HIVEP2 contains the ZAS domain comprised of two widely separated regions of zinc finger motifs, a stretch of highly acidic amino acids and a serine/threonine-rich sequence.
Specifications
| HIVEP2 | |
| Polyclonal | |
| Unconjugated | |
| HIVEP2 | |
| Agie-bp1; Angiotensinogen gene-inducible enhancer-binding protein 1; c-myc intron binding protein 1; DNA-binding protein AGIE-BP1; Hivep2; HIV-EP2; human immunodeficiency virus type 1 enhancer-binding protein 2; human immunodeficiency virus type I enhancer binding protein 2; Human immunodeficiency virus type I enhancer-binding protein 2; Human immunodeficiency virus type I enhancer-binding protein 2 homolog; MBP-2; MHC binding protein-2; MHC-binding protein 2; MIBP1; MIBP-1; MRD43; Myc intron-binding protein 1; Schnurri-2; SHN2; Shn-2; transcription factor HIVEP2; ZAS2; ZNF40B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 15273, 29721, 3097 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P31629, Q00900, Q3UHF7 | |
| HIVEP2 | |
| Synthesized peptide derived from human HIVEP2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction