Learn More
HJ 959-2018 Tetraethyl Lead and Isopropanol Mixture 490 100 μg/mL in Methanol, Dr. Ehrenstorfer
Specifications
Specifications
| CAS | 67-63-0, 78-00-2 |
| Quantity | 1 mL |
| Certifications/Compliance | ISO 17034 |
| Solvent or Matrix | Methanol |