Learn More
Invitrogen™ HK1 Recombinant Superclonal™ Antibody
Rabbit Recombinant Superclonal Antibody
Brand: Invitrogen™ 711777
Description
This antibody is predicted to react with Monkey, Horse, Dog, Bovine. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called Recombinant polyclonal antibody, this product is now rebranded as Recombinant Superclonal™ antibody. The physical product and the performance remain unchanged.
This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.
Specifications
| HK1 | |
| Recombinant Superclonal | |
| Unconjugated | |
| HK1 | |
| BB404130; brain form hexokinase; dea; downeast anemia; glycolytic enzyme; HEXOKIN; hexokinase; hexokinase 1; hexokinase I; hexokinase IR; hexokinase type I; Hexokinase, tumor isozyme; hexokinase-1; Hexokinase-A; HK; HK I; HK1; Hk-1; Hk1-s; HK1-ta; HK1-tb; HK1-tc; HKD; HKI; HMSNR; HXK1; mHk1-s | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3098 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P19367 | |
| HK1 | |
| Peptides corresponding to human HK1 [1) aa315-aa334 2) aa719-aa738]. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.