missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ HK3 Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA584306

Product Code. 16391255

  • € 586.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MDSIGSSGLR QGEETLSCSE EGLPGPSDSS ELVQECLQQF KVTRAQLQQI QASLLGSMEQ A Highest antigen sequence indentity to the following orthologs: Mouse - 69%, Rat - 67%.

Hexokinases phosphorylate glucose to produce glucose 6 phosphate, thus committing glucose to the glycolytic pathway. In vertebrates there are four major glucose phosphorylating isoenzymes, designated hexokinase I, II, III and IV (glucokinase). Similar to hexokinases I and II, the allosteric enzyme hexokinase III is inhibited by its product glucose 6 phosphate. The catalytic activity of hexokinase III is associated with the C terminus while regulatory function is associated with the N terminus.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

HK3
Polyclonal
Unconjugated
HK3
ATP:D-hexose 6-phosphotransferase; hexokinase 3; hexokinase 3 (white cell); hexokinase iii; hexokinase type III; Hexokinase3; Hexokinase-3; Hexokinase-C; HK III; HK3; HKIII; HK-III; HXK3; RNU73859
Rabbit
Antigen affinity chromatography
RUO
3101
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
P52790
HK3
Recombinant protein corresponding to Human HK3. Recombinant protein control fragment (Product #RP-103265).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.