missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA A Antibody (CATA-1), PE/Cy5.5, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47673PECY55
This item is not returnable.
View return policy
Description
HLA A Monoclonal specifically detects HLA A in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA A | |
| Monoclonal | |
| PE-Cyanine5.5 | |
| PBS with 0.05% Sodium Azide | |
| HLA-A | |
| Normal human peripheral blood lymphocytes of phenotype A1, Aw32, B7, B37, Cw-, Cw-, DR2, DRw10 | |
| 0.1 mL | |
| Primary | |
| This monoclonal antibody reacts with cells bearing HLA-A25 or HLA-Aw32 antigens. In addition, a reaction was observed with a cell of phenotype A2, Aw31; B17, Bw49. HLA-A, with HLA-B and HLA-C, belongs to major histocompatibility complex (MHC) class I antigens and expresses constitutively on all nucleated cells. HLA system comprises closely linked genes controlling highly polymorphic proteins involved in the presentation of peptides to the T-cell receptor, inhibition of NK cell cytotoxicity, and rejection of tissue allotransplantation. Specific alleles at HLA loci are associated with diseases. This monoclonal antibody is specifically applicable for typing peripheral T cells for the antigens HLA-A25 and HLA-Aw32. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| CATA-1 | |
| Flow Cytometry | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Mouse | |
| Protein A or G purified | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105.0 | |
| Human | |
| IgG2a κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido