missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA A2 Antibody (BB7.2), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-44896R
This item is not returnable.
View return policy
Description
HLA A2 Monoclonal specifically detects HLA A2 in Human samples. It is validated for Flow Cytometry.
Specifications
| HLA A2 | |
| Monoclonal | |
| DyLight 550 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| HLA-A | |
| HLA-A2 solubilised by papain | |
| Primary | |
| The antibody BB7.2 recognizes an epitope at the C-terminus of alpha-2 helix and a turn on one of the underlying beta strands within the human HLA-A2 histocompatibility antigen. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| BB7.2 | |
| Flow Cytometry | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Mouse | |
| 0.1 mL | |
| 3105 | |
| Human | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction