missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA ABC Antibody (MHC-I/8147R), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20573-100ug
This item is not returnable.
View return policy
Description
HLA ABC Monoclonal antibody specifically detects HLA ABC in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| HLA ABC | |
| Monoclonal | |
| 0.2 mg/ml | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A*1, MHC class I antigen A*11, MHC class I antigen A*80 | |
| Recombinant fragment (around aa100-300) of human HLA-I protein (exact sequence is proprietary) | |
| 100 μg | |
| Adaptive Immunity, Cell Biology, Immunology | |
| 3105 | |
| Store at 4&drg;C. Do not freeze. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| MHC-I/8147R | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction