missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HLA-ABCE Antibody (TP25.99SF), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68006CL1
This item is not returnable.
View return policy
Description
HLA-ABCE Monoclonal antibody specifically detects HLA-ABCE in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,Immunohistochemistry (Frozen)
Specifications
| HLA-ABCE | |
| Monoclonal | |
| CoraFluor 1 | |
| HLA-ABC, HLA-E, major histocompatibility complex, class I, A, major histocompatibility complex, class I, B, major histocompatibility complex, class I, C, major histocompatibility complex, class I, E, MHC class I | |
| IFN-gamma-treated human melanoma cells COLO 38. | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Western Blot, Immunohistochemistry (Frozen) | |
| TP25.99SF | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction